Tisk…
Pipette tips, universal, Axygen®
Dodavatel: Corning
MTX-1250-C MTX-1250CLR MTX-1250-C-R MTXF1250CRS T-1000-B T-1000-B-R T-1000-B-R-S T-1000-C T-1000-C-R T-1000-C-R-S T-1005-WB-C T1005WBCR T1005WBCRS T-200-C T-200-C-R T-200-C-R-S T-200-C-STK T-200-C-STKS T-200-Y T-200-Y-R T-200-Y-R-S T-200-Y-STK T-200-Y-STKS T-205-WB-C T-205-WB-C-R T-210-Y T-300 T-300-R T-300-R-S T-300-STK T-300-STK-S T-350-C T-350-C-R T-350-C-R-S T-400 T-400-R T-400-R-S TR-222-C TR-222CLSTKS TR-222-C-R TR-222-C-R-S TR-222-C-STK TR-222-Y TR-222-Y-R TR-222-Y-R-S TR-222-Y-STK TR-333-C-R TXL-10 TXL-10-R TXL-10-R-S T-1200-C-R-S T-5000-C TE-1004-B TE-1004-B-L-R TE-1004-B-R TE-204-Y-L-R TE-204-Y-L-R-S TE-204-Y-R TE-204-Y-R-S TE-204-Y T-205-WB-C-R-S TR-222-C-STK-S
613-1263EA 1810 CZK
613-1263 613-1265 613-1267 613-1269 613-0965 613-1270 613-1271 613-0669 613-0670 613-0630 613-1272 613-1276 613-1277 613-0649 613-0650 613-0629 613-0651 613-1288 613-0901 613-1289 613-1290 613-1291 613-1292 613-1293 613-1297 613-1299 613-0640 613-0641 613-0628 613-0642 613-1309 613-0666 613-0667 613-0668 613-0646 613-0647 613-0648 613-0656 613-3826 613-0657 613-0658 613-0659 613-3827 613-3828 613-3829 613-3830 613-0661 613-0643 613-0644 613-0645 613-0673 613-0663 613-1320 613-1334 613-1336 613-1340 613-1341 613-1342 613-1343 613-1338 615-0029 AXYGTR-222-C-STK-S
Pipette tips, universal, Axygen®
Špičky pipetovací
These universal pipette tips are compatible with most micropipettes.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Informace pro objednávání: Available loose or in racks, sterile or non sterile versions.