Tisk…
Multi-channel pipettes, electronic, variable volume, Finnpipette™ Novus
Dodavatel: Thermo Fisher Scientific
46300000 46300100 46300200 46300300 46300400 46300500 46300700 46300800
613-0176EA 0 CZK
613-0176 613-0177 613-0178 613-0179 613-0180 613-0181 613-0182 613-0612
Multi-channel pipettes, electronic, variable volume, Finnpipette™ Novus
Pipety Micropipettes
Ergonomic design for exceptionally easy use in routine laboratory work.
  • Handle can be pivoted through 120° so that pipetting can be controlled with the index finger
  • Soft-touch eject button suitable for right- or left handed use
  • Large screen, two level menu can be accessed with a single button
  • Ten pipetting functions and nine speed levels accessible via a single button, enables fast programming
  • Autoclavable (end fitting only)
Informace pro objednávání: Supplied with a universal AC adapter, a complete operating and maintenance manual, a certificateMore Product Information