Tisk…
VWR® Burettes, with Detachable PTFE Stopcock
Dodavatel: VWR Collection
612-6694EA 1660 CZK
612-6694 612-6695 612-6696
VWR® Burettes, with Detachable PTFE Stopcock
Byrety
Borosilicate 3.3 glass, class AS.
  • High chemical and thermal resistance
  • Schellbach stripe for accurate meniscus reading
  • Detachable and interchangeable PTFE stopcock
  • High contrast blue enamel graduations
  • Batch certificate included
No breakage in case of improper handling because the stopcock is not directly connected with the body of the burette. Fast to dismantle, easy to clean and simple to repair because all individual components are replaceable.
Certifikace: DIN EN ISO 385