Tisk…
Mixing Measuring cylinders, tall form, class B, borosilicate glass
Dodavatel: witeg Labortechnik
3.752.001 3.752.005 3.752.010 3.752.025 3.752.050 3.752.100 3.752.003
612-4960EA 432 CZK
612-4960 612-4961 612-4962 612-4963 612-4964 612-4965 612-4966
Mixing Measuring cylinders, tall form, class B, borosilicate glass
Válce Válce odměrné
Borosilicate glass, class B, with PE stopper.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, NS ground socket
  • Brown graduations
Certifikace: DIN EN ISO 4788