Tisk…
Pipette tray for drawer
Dodavatel: KARTELL
252
612-1805EA 1830 CZK
612-1805
Pipette tray for drawer
Podnosy/Misky/Kelímky Podnosy na pipety
PVC, white.
  • Designed to hold about 30 pipettes
  • Four compartments to accommodate 1, 2, 5, 10 ml pipettes
W×D×H: 425×300×30 mm.