Tisk…
Pipette jars, with lid, Nalgene®
Dodavatel: Thermo Fisher Scientific
5242-0040 5242-0050 5242-0060
612-1803EA 7320 CZK
612-1803 612-1804 612-1827
Pipette jars, with lid, Nalgene®
Nádoby Válce na pipety
Made of cross-linked high-density polyethylene, with PPCO base.
  • Tough, stiff, seamless jars with dust cover resist stress-cracking
  • Ideal for soaking glassware, washing pipets with corresponding Nalgene® rinsers and baskets
  • Flanged base for added stability
Please note: The formula shown on the jar's label is effective and lasts longer than standard solutions. Do not mix this solution in the pipet jar. Keep jar on a drainboard, in a sink or in a container large enough to retain jar contents in case of leakage.
Upozornení: Concentrated sulfuric acidMore Product Information