Tisk…
Dishes, disposable
Dodavatel: Korff
60505 60530 60540 60545 60550 60560 60558 60500 60510 60520 60525 60552
KORF60505EA 1450 CZK
KORF60505 KORF60530 KORF60540 KORF60545 KORF60550 KORF60560 KORF60558 611-9031 611-9032 611-9033 611-9034 611-9035
Dishes, disposable
Lodičky Lodičky navažovací
Aluminium.
  • Lightweight
  • Good chemical resistance
  • Good temperature resistance
Weighing/drying pan, oil-free, ideal for general purpose weighing, dispensing, storage and drying.