Tisk…
Cubis® II High-Capacity Micro Balances
Dodavatel: Sartorius Balances
MCA116S3CE MCA116S3CEH MCA116S3CE1 MCA116S3CE1H MCA116S3CE1I MCA116S3CE1M MCA116S3CE99 MCA116S3CE99H MCA116S3S00 MCA116S3S00H MCA116S3S00I MCA116S3S00M MCA116S3S001 MCA116S3S001H MCA116S3S001I MCA116S3S001M MCA116S3S0099 MCA116S3S0099H MCA116S3S0099I MCA116S3S0099M MCA36P3CE MCA36P3CEH MCA36P3CE1 MCA36P3CE1H MCA36P3CE99 MCA36P3CE99H MCA36P3S00 MCA36P3S00H MCA36P3S00I MCA36P3S00M MCA36P3S001 MCA36P3S001H MCA36P3S001I MCA36P3S001M MCA36P3S0099 MCA36P3S0099H MCA36P3S0099I MCA36P3S0099M MCA36S3CE MCA36S3CEH MCA36S3CE1 MCA36S3CE1H MCA36S3CE99 MCA36S3CE99H MCA36S3CE99I MCA36S3CE99M MCA36S3S00 MCA36S3S00H MCA36S3S00I MCA36S3S00M MCA36S3S001 MCA36S3S001H MCA36S3S001I MCA36S3S001M MCA36S3S0099 MCA36S3S0099H MCA36S3S0099I MCA36S3S0099M MCA66P3CE MCA66P3CEH MCA66P3CE1 MCA66P3CE1H MCA66P3CE99 MCA66P3CE99H MCA66P3S00 MCA66P3S00H MCA66P3S00I MCA66P3S00M MCA66P3S001 MCA66P3S001H MCA66P3S001I MCA66P3S001M MCA66P3S0099 MCA66P3S0099H MCA66P3S0099I MCA66P3S0099M MCA66S3CE MCA66S3CEH MCA66S3CE1 MCA66S3CE1H MCA66S3CE99 MCA66S3CE99H MCA66S3S00 MCA66S3S00H MCA66S3S00I MCA66S3S00M MCA66S3S001 MCA66S3S001H MCA66S3S001I MCA66S3S001M MCA66S3S0099 MCA66S3S0099H MCA66S3S0099I MCA66S3S0099M MCE36P3CE MCE36P3CEH MCE36P3CEI MCE36P3CEM MCE36P3CF MCE36P3CFH MCE36P3CFI MCE36P3CFM MCE36P3S00 MCE36P3S00H MCE36P-3S00-D MCE36P3S00M MCE36S3CE MCE36S3CEH MCE36S3CEI MCE36S3CEM MCE36S3CF MCE36S3CFH MCE36S3CFI MCE36S3CFM MCE36S3S00 MCE36S3S00H MCE36S3S00I MCE36S3S00M MCE66P3CE MCE66P3CEH MCE66P3CEI MCE66P3CEM MCE66P3CF MCE66P3CFH MCE66P3CFI MCE66P3CFM MCE66P3S00 MCE66P3S00H MCE66P3S00I MCE66P3S00M MCE66S3CE MCE66S3CEH MCE66S3CEI MCE66S3CEM MCE66S3CF MCE66S3CFH MCE66S3CFI MCE66S3CFM MCE66S3S00 MCE66S3S00H MCE66S3S00I MCE66S3S00M MCA66S3CEUDQP1
611-5540EA 885890 CZK
611-5540 611-5539 611-5552 611-5550 611-5551 611-5549 611-5564 611-5563 611-5490 611-5488 611-5489 611-5487 611-5510 611-5508 611-5509 611-5507 611-5530 611-5528 611-5529 611-5527 611-5534 611-5533 611-5544 611-5543 611-5558 611-5557 611-5478 611-5476 611-5477 611-5475 611-5498 611-5496 611-5497 611-5495 611-5518 611-5516 611-5517 611-5515 611-5532 611-5531 611-5542 611-5541 611-5556 611-5554 611-5555 611-5553 611-5474 611-5472 611-5473 611-5471 611-5494 611-5492 611-5493 611-5491 611-5514 611-5512 611-5513 611-5511 611-5538 611-5537 611-5548 611-5547 611-5562 611-5561 611-5486 611-5484 611-5485 611-5483 611-5506 611-5504 611-5505 611-5503 611-5526 611-5524 611-5525 611-5523 611-5536 611-5535 611-5546 611-5545 611-5560 611-5559 611-5482 611-5480 611-5481 611-5479 611-5502 611-5500 611-5501 611-5499 611-5522 611-5520 611-5521 611-5519 611-5446 611-5444 611-5445 611-5443 611-5462 611-5460 611-5461 611-5459 611-5298 611-5295 611-5296 611-5294 611-5442 611-5440 611-5441 611-5439 611-5458 611-5456 611-5457 611-5455 611-5293 611-5291 611-5292 611-5290 611-5454 611-5452 611-5453 611-5451 611-5470 611-5468 611-5469 611-5467 611-5422 611-5420 611-5421 611-5419 611-5450 611-5448 611-5449 611-5447 611-5466 611-5464 611-5465 611-5463 611-5418 611-5416 611-5417 611-5415 SARBMCA66S3CEUDQP1
Cubis® II High-Capacity Micro Balances
Váhy
The Cubis® II laboratory balances are modular, providing choice between applications and configurations that best suit the users' needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions. The Cubis® II range of high-capacity micro balances with a maximum load between 32 and 111 g and a readability between 0,001 mg and 0,002 mg provide the ideal choice for a broad range of applications.
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity
  • 21 CFR Part 1
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Typ přípojky IEC plug IEC plug IEC plug
Napětí 100 - 240 V 100 - 240 V 100 - 240 V