Tisk…
VWR®, Platform Scales
Dodavatel: VWR Collection
611-5085EA 26100 CZK
611-5085 611-5074 611-5069 611-5071 611-5075 611-5078 611-5079 611-5077 611-5080 611-5086 611-5081 611-5088
VWR®, Platform Scales
Váhy
Industrial series of advanced platform scales, offering increased accuracy and fast stabilisation time, combined with easy, intuitive operation and a large graphical screen for laboratory applications and industrial settings. Ideal for weighing, mixing, counting and check weighing.
  • Efficient filtering algorithms to deliver quick and accurate results
  • Fast stabilisation time
  • Easy to clean and high chemical resistance
  • Choice of two platform sizes (M-Line 280×280 mm, L-Line 300×400 mm)
  • Different column size, for tabletop or ground weighing (M-
More Product Information