Tisk…
Přesné váhy, řada Entris II Advanced
Dodavatel: Sartorius Balances
BCA10201-1S BCA10201I-1S BCA1202-1S BCA1202I-1S BCA1203I-1S BCA12201-1S BCA12201I-1S BCA2201-1S BCA2201I-1S BCA2202-1S BCA2202I-1S BCA223-1S BCA223I-1S BCA3202-1S BCA3202I-1S BCA323-1S BCA323I-1S BCA4202-1S BCA4202I-1S BCA423-1S BCA423I-1S BCA5201-1S BCA5201I-1S BCA6202-1S BCA6202I-1S BCA623-1S BCA623I-1S BCA8201-1S BCA8201I-1S BCA822-1S BCA822I-1S BCA10201I-1CEU BCA1202I-1CEU BCA1203I-1CEU BCA12201I-1CEU BCA2201I-1CEU BCA2202I-1CEU BCA223I-1CEU BCA3202I-1CEU BCA323I-1CEU BCA4202I-1CEU BCA423I-1CEU BCA5201I-1CEU BCA6202I-1CEU BCA623I-1CEU BCA8201I-1CEU BCA822I-1CEU
611-4837EA 0 CZK
611-4837 611-4838 611-4839 611-4840 611-4841 611-4842 611-4843 611-4846 611-4847 611-4848 611-4849 611-4850 611-4851 611-4854 611-4855 611-4856 611-4857 611-4859 611-4860 611-4861 611-4862 611-4863 611-4864 611-4865 611-4866 611-4867 611-4868 611-4871 611-4872 611-4873 611-4874 611-4875 611-4877 611-4879 611-4881 611-4885 611-4887 611-4889 611-4893 611-4895 611-4899 611-4901 611-4903 611-4905 611-4907 611-4911 611-4913
Přesné váhy, řada Entris II Advanced
Váhy
Entris® II Advanced precision balances feature isoCAL, intuitive graphic touch display and 13 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard wearing PBT, stainless steel and glass. Integrated protection systems increase reliability of weighing results: Three configurable levels determine valid weighing data and ensure only valid data is transferred to external devices.
  • Fast stabilisation time thanks to state-of-the-art weighing sensors
More Product Information