Tisk…
VWR®, Digital Timer, Four Channels
Dodavatel: VWR Collection
609-0181EA 1720 CZK
609-0181
VWR®, Digital Timer, Four Channels
Časovace/Stopky Časovače digitální
This 4-channel timer has a capacity of 99 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
  • Counts either up or down
  • Remembers the last time set
  • Lay flat on a bench, use the flip-open stand for a vertical view, or carry it on a lab coat with the convenient pocket clip
Certifikace: Featuring a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipmentMore Product Information