Tisk…
Anti-Ly-6G Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7) [clone: 1A8]
Dodavatel: Tonbo Biosciences
60-1276-U025 60-1276-U100
60-1276-U025EA 1840 CZK
60-1276-U025 60-1276-U100
Anti-Ly-6G Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7) [clone: 1A8]
Protilátky
The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.
In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: forMore Product Information