Tisk…
Suction filter, b.safe
Dodavatel: Bohlender
M515-01
590-0201EA 1940 CZK
590-0201
Suction filter, b.safe
Filtry Filtry pro práci s kapalinami
Filter candle made of porous PE.
  • With a bore to be put on capillary tubing
  • Restrains pollution from solvents
  • For constant analysis results
Exchange the filter regularly every three months for reliable operation.