Tisk…

Ammonia solution 29%, Finyte® Microelectronic, J.T.Baker®

Dodavatel: Avantor

Synonyms: Vodný roztok amonia, Hydroxid amonný, Amoniak

5820-15EA 7360 CZK
5820-15
Ammonia solution 29%, Finyte® Microelectronic, J.T.Baker®
Hydroxid amonný
Vzorec: NH₃
MW: 35.05 g/mol
MDL Number: MFCD00066650
CAS číslo: 1336-21-6
UN: 2672
ADR: 8 III

Objednat

Výsledky specifikačního testu

For Microelectronic Use
Appearance Passes Test
Assay (as NH₃) 28.0 - 30.0 %
Color (APHA) ≤10
Residue after Ignition ≤3 ppm
Carbon Dioxide (CO₂) ≤10 ppm
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤0.2 ppm
Nitrate (NO₃) ≤0.4 ppm
Phosphate (PO₄) ≤0.10 ppm
Total Sulfur (as SO₄) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤5.0 ppb
Trace Impurities - Calcium (Ca) ≤20 ppb
Trace Impurities - Chromium (Cr) ≤5.0 ppb
Trace Impurities - Cobalt (Co) ≤1.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤5.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silicon (Si) ≤10.0 ppb
Trace Impurities - Silver (Ag) ≤5.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and greater ≤40 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR