Tisk…
Gas purifiers, GateKeeper® GPU 35 4R
Dodavatel: ENTEGRIS
GPUS35FIX04R00 GPUS35FHX04R00
568-0920EA 0 CZK
568-0920 568-0918
Gas purifiers, GateKeeper® GPU 35 4R
Filtry Filtry na plyny
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 35 is designed for peak flow rates up to 1 slpm.
Certifikace: CEMore Product Information