Tisk…
Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Dodavatel: Brady
306883 306886 306889 306880
568-0897EA 86360 CZK
568-0897 568-0899 568-0901 568-0895
Industrial label printers with Brady workstation LAB suite, BradyPrinter i7100
Tiskárny Label Printers
The BradyPrinter i7100 industrial thermal transfer bench top printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and lab markets.
  • 300 or 600 dpi
  • Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
  • Mono colour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. TheMore Product Information