Tisk…
Capillary columns, BPX5
Dodavatel: Trajan Scientific and Medical
054099 054100 054101 054102 0541025 054104 054112 054113 054114 054118 054119 0541205 054121 054122 054123 054125 054126 054127 054129 054130 054133 054134 0541345 0541348 054142 054144 054145 054146 054148 054153 054154 054160 054110 0541261 0542170 0541011
568-0101EA 12360 CZK
568-0101 558-0319 558-0248 568-0102 568-0103 558-0111 558-0116 558-0117 558-0118 568-0106 568-0107 568-0109 568-0110 568-0111 568-0112 558-0123 558-0124 558-0125 558-0127 568-0114 558-0320 568-0117 568-0119 568-0121 558-0129 558-0131 558-0132 558-0133 558-0135 558-0321 568-0126 568-0129 568-0105 568-0113 568-0000 568-0368
Capillary columns, BPX5
Kolony chromatografické
These columns are suitable for trace analysis of pesticides, drugs, hydrocarbons and phenols. The BPX5 phase has been designed for robustness and is suitable for over 80% of all routine analyses performed by chromatographers.
5% phenyl polysilphenylene-siloxane.
Max. temperature: 370 °C
Suitable replacement for: CP-Sil 8CB, DB-5, DB-5.625 XTI-5, DB-5MS, HP-5, HP-5MS, HP5-TA, MDN-5S, Rtx-5ms, SPB-5, Ultra-2, VB-5, ZB-5, VF-5MS.