Tisk…
Discovery® HS F5 PFP HPLC Columns
Dodavatel: Merck
567500-U 567501-U 567502-U 567503-U 567504-U 567506-U 567507-U 567508-U 567510-U 567511-U 567512-U 567513-U 567515-U 567516-U 567517-U 567520-U 567523-U 567530-U 567531-U 567532-U 567535-U 567536-U 567542-U 567570-U 567571-U 567572-U 567573-U 567574-U 567575-U 567576-U 567577-U
567500-UEA 18190 CZK
567500-U 567501-U 567502-U 567503-U 567504-U 567506-U 567507-U 567508-U 567510-U 567511-U 567512-U 567513-U 567515-U 567516-U 567517-U 567520-U 567523-U 567530-U 567531-U 567532-U 567535-U 567536-U 567542-U 567570-U 567571-U 567572-U 567573-U 567574-U 567575-U 567576-U 567577-U
Discovery® HS F5 PFP HPLC Columns
Kolony chromatografické
The Discovery® HS F5 (PFP) HPLC Column provides excellent peak shape and stable, low-bleed LC-MS separation.
  • Pentafluorophenyl terminated reversed phase column with unique retention and selectivity (basic and halogenated compounds)
  • HS F5 bonded phase provides reversed phase separations that are distinctly different from C18 columns
  • Compounds will generally elue within the same retention tie window, making most C18 methods easily transferable
  • Impurity: <10 ppm metals
  • Surface coverage: 4 µmol/m²
Designed to meet the exacting requirements ofMore Product Information