Tisk…
Hydrogen generator, Precision Hydrogen Trace
Dodavatel: PEAK SCIENTIFIC
64-0500 64-0250
564-0173EA 389000 CZK
564-0173 564-0300
Hydrogen generator, Precision Hydrogen Trace
Generátory plynu Generátory vodíku
This generator is suitable for carrier gas and flame gas at trace detection limits. Hydrogen purity 99,9999%. Moisture content <1 ppm.
  • Proven PEM Technology generates hydrogen safely and reliably
  • Regenerative PSA Dryers ensure highest level of purity
  • Automatic loading pump as standard
  • Maintenance limited to replacing de-ionizer cartridge
  • Internal leak detection with automatic shutdown features
  • Series option to combine multiple units for higher flow requirements
  • Remote shutdown
  • GC In Oven Hydrogen Leak Detector available as an optional extra