Tisk…
Nitrogen and dry air gas generator, Genius 3045
Dodavatel: PEAK SCIENTIFIC
10-3045
564-0280EA 769430 CZK
564-0280
Nitrogen and dry air gas generator, Genius 3045
Generátory plynu Generátory dusíku
Designed to the specifications of the Bruker EVOQ triple Quadrupole's gas requirements delivering dry air at up to 50 L/min and nitrogen at up to 32 L/min.
  • Independent nitrogen and dry air gas supplies can be tailored for its application
  • Higher flow 30 series generator designed for hungrier LC-MS applications
  • Compressor based solution, no need for an external air supply
  • Highly economical source of nitrogen gas with low lifetime running costs
  • Gas is supplied on demand so generator works to your schedule
  • Service indication to allow planning of preventative
More Product Information