Tisk…

Hydroxid sodný 0.5 N, BAKER ANALYZED® ACS volumetric solution, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Caustic soda

5634-02EA 1880 CZK
5634-02
Hydroxid sodný 0.5 N, BAKER ANALYZED® ACS volumetric solution, J.T.Baker®
Hydroxid sodný

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to +/- 1 part per 1000, provided on label.

Vzorec: NaOH
MW: 40
Bod varu: 1390
Bod tání: 323
Hustota: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS číslo: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use 0.499 - 0.501
Protect from air to avoid absorption of carbon dioxide. ≤5 ppm
Normality 0.499 - 0.501
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR