Tisk…

jod 0.05 mol/l (0.1N), BAKER ANALYZED® Volumetric, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

5623.1000EA 1700 CZK
5623.1000
jod 0.05 mol/l (0.1N), BAKER ANALYZED® Volumetric, J.T.Baker®
jod

BAKER ANALYZED™ Reagent—A grade of high-quality chemicals for laboratory and specialized industrial use.

Vzorec: I₂
MW: 253.81
Bod varu: 184.3
Bod tání: 114
MDL Number: MFCD00011355
CAS číslo: 7553-56-2
Merck Index: 13,05036

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use 0.0995 - 0.1005
Normality 0.0995 - 0.1005
Standardization at 25°C traceable to NIST Standard Reference Material.

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR