Tisk…

Aceton, Selectipur® for the electronics industry

Dodavatel: ELECTRONIC CHEMICALS
Nebezpečí

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

55954791
55954791.EA 0 CZK
55954791.
Aceton, Selectipur® for the electronics industry
Aceton

Product from BASF


Informace o dodávce: This product is not available in all countries. Please check with your local VWR office or supplier.


Upozornení: This product is not available in all countries. Please check with your local VWR office or supplier.

Vzorec: CH₃COCH₃
Bod varu: 56,2 °C (1013 hPa)
Bod tání: –95,4 °C
Hustota: 0,792 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS číslo: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR