Tisk…
Capillary columns, DB-WAX
Dodavatel: Agilent
122-7032 123-7032 123-7033 125-7032 125-7037 128-7032 123-7032E 128-7022 128-7052 121-7012LTM 121-7022E 121-7022LTM 121-7023LTM 121-7042 121-7042E 122-7012LTM 122-7013LTM 122-7032LTM 122-7033LTM 123-7012LTM 123-7013LTM 123-7032LTM 123-7033LTM 125-7031LTM 125-7032LTM 127-7012LTM 127-7013LTM 127-7022LTM 127-7023LTM 128-7032LTM 222-7013LTM 222-7033LTM 122-7032UI
558-0367EA 0 CZK
558-0367 HEWL123-7032 558-0370 558-0374 HEWL125-7037 HEWL128-7032 HEWL123-7032E HEWL128-7022 HEWL128-7052 HEWL121-7012LTM HEWL121-7022E HEWL121-7022LTM HEWL121-7023LTM HEWL121-7042 HEWL121-7042E HEWL122-7012LTM HEWL122-7013LTM HEWL122-7032LTM HEWL122-7033LTM HEWL123-7012LTM HEWL123-7013LTM HEWL123-7032LTM HEWL123-7033LTM HEWL125-7031LTM HEWL125-7032LTM HEWL127-7012LTM HEWL127-7013LTM HEWL127-7022LTM HEWL127-7023LTM HEWL128-7032LTM HEWL222-7013LTM HEWL222-7033LTM HEWL122-7032UI
Capillary columns, DB-WAX
Kolony chromatografické
These columns improve resolution of low boiling point analytes.
  • High polarity
  • Lower temperature limit of 20 °C
  • Column-to-column reproducibility
  • Bonded and cross-linked
  • Solvent rinsable
Polyethylene glycol (PEG)