Tisk…
Aqueous/organic size exclusion chromatography columns, MSpak GF-310
Dodavatel: Resonac
F7600110
556-0063EA 59090 CZK
556-0063
Aqueous/organic size exclusion chromatography columns, MSpak GF-310
Kolony chromatografické
Semi-micro columns for Asahipak GF-310 HQ. Effective for drug analysis in biological samples by using the column-switching method to remove proteins.
  • Useful for LC-MS analysis
  • Suitable for the analysis of surfactants