Tisk…
BioHPLC column, DNASwift™ SAX-1S
Dodavatel: Thermo Fisher Scientific
066766
555-7161EA 46370 CZK
555-7161
BioHPLC column, DNASwift™ SAX-1S
Kolony chromatografické
This column is a strong anion-exchange column that provides exceptionally high resolution for the purification of oligonucleotides.
  • Compatible with high pH, solvents and high temperatures
  • Pressure and chemically stable monolith support coated with functionalised latex nanobeads