Tisk…
Ion exchange HPLC columns, monolith, ProSwift™ IEX
Dodavatel: Thermo Fisher Scientific
064293 064294 064295 066765
555-7225EA 43040 CZK
555-7225 555-7141 555-7142 555-7160
Ion exchange HPLC columns, monolith, ProSwift™ IEX
Kolony chromatografické
High resolution columns used for fast protein analysis.
  • High loading capacity
  • Wide range of operational flow rates
  • Reproducibility and ruggedness
  • High throughput and improved productivity
These monolith ion-exchange columns deliver the outstanding resolving power of nonporous media combined with the high capacity of conventional porous media. They offer excellent separations using fast gradients at low and high flow rates, which improve productivity.