Tisk…
HPLC columns, Hypersil™ Silica
Dodavatel: Thermo Fisher Scientific
30005-052130 30005-014001 30003-014001 30005-154030 30005-054630 30005-254630 30005-154630 30005-254030 30005-102130 30005-054030 30003-154630 30003-154030
555-6300EA 24070 CZK
555-6300 555-6297 555-6335 555-6312 555-6303 555-6319 555-6313 555-6318 555-6305 555-6302 555-6350 555-6349
HPLC columns, Hypersil™ Silica
Kolony chromatografické
Hypersil™ Silica columns are suitable for normal phase chromatography of non-polar and moderately polar organic compounds.
  • Good batch-to-batch reproducibility
  • Spherical particle with narrow particle size distribution
  • Long column lifetime
  • High performance and column efficiency