Tisk…
HPLC columns, NUCLEODUR® C18 Gravity-SB
Dodavatel: MACHEREY-NAGEL
760593.20 760616.20 760616.30 760616.40 760616.46 760618.20 760618.30 760618.40 760618.46 760619.20 760619.30 760619.40 760619.46 761992.20 761992.30 760591.20 760591.30 760591.40 760591.46 760593.30 760593.40 760593.46 760595.20 760596.20 760596.30 760596.40 760596.46 760598.20 760603.20 760603.30 760603.40 760603.46 760605.46 760606.20 760606.30 760606.40 760606.46 760607.20 760607.30 760607.40 760607.46 760608.20 760608.30 760608.40 760608.46 760609.20 760609.30 760609.40 760609.46 760613.20 760613.30 760613.40 760613.46 760615.46 760617.20 760617.30 760617.40 760617.46 761990.20 761990.30 761991.20 761991.30
MANA760593.20EA 22050 CZK
MANA760593.20 554-5823 554-5824 554-5825 554-5826 554-5827 554-5828 554-5829 554-5830 554-5831 554-5832 554-5833 554-5834 554-5835 554-5836 MANA760591.20 MANA760591.30 MANA760591.40 MANA760591.46 MANA760593.30 MANA760593.40 MANA760593.46 MANA760595.20 MANA760596.20 MANA760596.30 MANA760596.40 MANA760596.46 MANA760598.20 MANA760603.20 MANA760603.30 MANA760603.40 MANA760603.46 MANA760605.46 MANA760606.20 MANA760606.30 MANA760606.40 MANA760606.46 MANA760607.20 MANA760607.30 MANA760607.40 MANA760607.46 MANA760608.20 MANA760608.30 MANA760608.40 MANA760608.46 MANA760609.20 MANA760609.30 MANA760609.40 MANA760609.46 MANA760613.20 MANA760613.30 MANA760613.40 MANA760613.46 MANA760615.46 MANA760617.20 MANA760617.30 MANA760617.40 MANA760617.46 MANA761990.20 MANA761990.30 MANA761991.20 MANA761991.30
HPLC columns, NUCLEODUR® C18 Gravity-SB
Kolony chromatografické
NUCLEODUR® C18 Gravity-SB is a monomeric octadecyl modified silica phase with extensive endcapping for HPLC.
  • High purity spherical silica gel
  • Hydrophobic phase with distinct polar selectivity
  • Suitable for LC/MS due to low bleeding characteristics
C18 Gravity-SB with its additional distinct polar selectivity is ideal for method development. Compared to monomeric octadecyl phases with conventional endcapping this premium phase shows better retention of early eluting substances. Thus, it is successfully applied for sophisticated analytical separations,More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Naplnění uhlíkem 13%
S navazanou skupinou Yes
Rozsah pH 1 - 9
Kód USP L1