Tisk…
SafetyCaps
Dodavatel: S.C.A.T.
107035 107607 107030 107031 107032 107058 107105 107637 107742 107512 107511 307019 307909 307910 307410 307520 399019 399909 307006 307100 307101
590-0521EA 7740 CZK
590-0521 590-0553 590-0516 590-0517 590-0518 221-0262 554-3494 SCTV107637 SCTV107742 554-4688 SCTV107511 SCTV307019 SCTV307909 SCTV307910 SCTV307410 SCTV307520 SCTV399019 SCTV399909 SCTV307006 SCTV307100 SCTV307101
SafetyCaps
Uzávěry Uzávěry bezpečnostní a uzávěry bezpečnostní na odpad
The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the
More Product Information