Tisk…

Anion exchange chromatography columns, IEC series

Dodavatel: Resonac
F6110011 F6118255
554-2853EA 30670 CZK
554-2853 554-2857
Anion exchange chromatography columns, IEC series
Kolony chromatografické
QA-825, DEAE-825: Suitable for the analysis of relatively high molecular weight compounds, such as proteins, peptides, DNA, and RNA. Usable across wide pH range between 2 and 12.

DEAE3N-4T: Non porous base material. For rapid analysis.
Objednat

Produktová specifikace

Environmentally Preferable
Maximální tlak 2,0 MPa (20 bar)
Rozsah pH 2 - 12
Teplotní limit 10 - 50 °C
Kód USP L23

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR