Tisk…
HPTLC Vario system
Dodavatel: CAMAG
022.8550
552-0049EA 72920 CZK
552-0049
HPTLC Vario system
TLC monitoring desek
Time saving by optimisation of development conditions.
  • Development with six different solvents can be tested side by side
  • Both sandwich and tank configurations can be simulated side by side, making results directly comparable
  • Six different conditions of pre-equilibration, including relative humidity, can be tested simultaneously
  • These variations of developing conditions can be freely combined
Informace o dodávce: The HPTLC Vario system comprises an HPTLC Vario chamber for 100×100 mm plates and an HPTLC scoring unit for preparation of TLC/HPTLC plates.