Tisk…
Nanomat 4, TLC plate applicator
Dodavatel: CAMAG
022.4730
552-0007EA 35240 CZK
552-0007
Nanomat 4, TLC plate applicator
TLC aplikátory
The Nanomat 4 is used for easy application of samples in the form of spots onto TLC and HPTLC layers. A notch system ensures that sample positioning meets the requirements for automatic scanning with a densitometer. Samples are dispensed in volumes of 0,5 to 5 µl with disposable capillaries loaded into the universal capillary holder (552-0044) using the capillary guide (552-0039).
  • Easy to use and convenient
  • Fast, accurate reproducible spotting
  • Suitable for TLC and HPTLC plates