Tisk…
Gastight® 1700 series syringes
Dodavatel: HAMILTON BONADUZ
201000 201050 201100 201150 201500 201600 202134 80000 80001 80008 80030 80039 80075 80200 80201 80208 80230 80239 80900 80901 80908 80920 80922 80930 80939 81000 81001 81008 81020 81022 81030 81039 81100 81101 81120 81122 81130 81139 81175 81201 81217 81220 81222 81230 81243 81227 7656-01 7655-01 7657-01 203566 7653-01 81127 7654-01 80011 80927 81026 81027 0159902 0160310 203560 203563 7658-01 80266 81041 81242 CAL80000 CAL80200 CAL80900 CAL81000 CAL81100 CAL81217 CAL81227 CAL7658-01 CAL81030 CAL81020 80085 80016 CAL81230 208639 CAL81220 208637 208641 208643 CAL80239 81085 81185 80285 80985
548-2261EA 3510 CZK
548-2261 548-2262 613-1604 548-2263 613-0105 613-1119 HAMI202134 549-1124 549-1125 548-1962 549-1126 548-2286 549-0220 549-1130 549-1131 548-2288 549-1132 548-2289 549-1169 549-1170 548-1964 613-0496 613-1421 549-1171 549-0364 549-1174 549-1175 548-2309 549-0365 613-1422 549-1176 549-0366 549-1179 549-1180 549-0367 613-1423 549-1181 548-1965 549-1183 549-1184 549-1186 613-1425 613-1424 549-1187 549-0368 548-2313 548-2281 548-1961 548-2282 HAMI203566 548-1960 548-2312 548-2280 548-2284 548-2308 613-4419 548-2310 548-2259 548-2260 548-2266 HAMI203563 548-2283 548-2290 548-2311 548-2314 548-2330 548-2331 548-2338 548-2339 548-2341 548-2343 548-2344 548-2329 548-2340 548-1974 HAMI80085 548-2285 HAMICAL81230 548-2268 548-1975 548-2267 548-2269 548-2270 HAMICAL80239 HAMI81085 HAMI81185 HAMI80285 HAMI80985
Gastight® 1700 series syringes
Stříkačky Stříkačky chromatografické
These syringes are the Gastight® version of the original 700 series syringes and are ideal for dispensing volumes from 1 µl up to 500 µl.
  • Fluid path includes stainless steel, borosilicate glass and PTFE
  • Gas sterilisable
Gastight® syringes are ideal for dispensing both liquids and gases and have a precision machined PTFE plunger tip which creates a leak free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that aMore Product Information