Tisk…
Syringes, Neuros™
Dodavatel: HAMILTON BONADUZ
65460-15 65457-01 65458-01 65459-01 65460-02 65460-05 65460-10 65460-20 65457-02 65458-02 65459-02 65460-03 65460-06 65460-11 65460-16 65460-21
549-0703EA 11230 CZK
549-0703 549-0697 549-0698 549-0699 549-0700 549-0701 549-0702 549-0704 HAMI65457-02 HAMI65458-02 HAMI65459-02 HAMI65460-03 HAMI65460-06 HAMI65460-11 HAMI65460-16 HAMI65460-21
Syringes, Neuros™
Stříkačky Stříkačky chromatografické
Developed specifically for the neurosciences, these syringes minimise injection site damage by increasing needle rigidity and delivering a micro volume of liquid to an exact location.
  • Compatibility with most infusion pumps and stereotaxic holders
  • Black needle sleeve indicates the exposed needle length
The technology is offered on several models from the 700 Microliter™, 1700 Gastight® and 7000 Modified Microliter™ series. These syringes have very little to no dead volume which preserves the maximum amount of buffer and solution. They feature twoMore Product Information