Tisk…
Vial racks, WHEATON®
Dodavatel: DWK Life Sciences
868804 868808 868810 985750
548-2157EA 5010 CZK
548-2157 548-1976 548-2158 548-2156
Vial racks, WHEATON®
Stojany Stojany na chromatografické vialky
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
  • Autoclavable and stackable
  • Good chemical resistance
  • Includes position identifiers