Tisk…
Headspace crimp vials, Chromacol™
Dodavatel: CHROMACOL
9-CV
548-1094EA 2370 CZK
548-1094
Headspace crimp vials, Chromacol™
Vialky
Clear borosilicate glass vials.
  • Bevelled edge, 20 mm crimp finish
  • Round bottom (with exception of 27-CV which is flat)