Tisk…
VWR®, Crimp Neck Vials ND11
Dodavatel: VWR Collection
548-9006AEA 7760 CZK
548-9006A 548-0423A 548-0172A 548-2500A 548-0905A 548-3311A 548-1861A 548-0467A 548-0173A 548-0174A 548-1858A 548-0003A 548-0447A 548-0004A 548-0005A 548-3337A 548-3310A 548-8008A 548-8009A 548-8010A 548-2380A 548-2148A 548-1361A 548-0171A 548-2159A 548-1362A 548-0856A 548-0474A 548-1465A 548-1364A 548-1400A 548-0906A 548-3254A 548-0092A 548-2870A 548-1516A 548-2126A 548-0418A 548-0421A 548-1840A 548-2146A 548-2362A 548-2512A 548-2603A 548-2604A
VWR®, Crimp Neck Vials ND11
Vialky
ND11 crimp neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Crimp top vials provide an excellent seal
  • Larger opening improves sample accessibility
ND11 crimp neck vials are available in amber or clear version. For use in a broad variety of instruments as GC and HPLC standard vials with a flat bottom. Multiple crimp cap colours mostly available with assembled stopper or septa are engineered toMore Product Information