Tisk…

n-Heptan, ChromAR® for HPLC, Macron Fine Chemicals™

Dodavatel: Avantor Performance Materials

Synonyms: high purity solvents

5410-25EA 6970 CZK
5410-25
n-Heptan, ChromAR® for HPLC, Macron Fine Chemicals™
n-Heptane
Vzorec: H₃C(CH₂)₅CH₃
MW: 100.2
Bod varu: 98.4
Bod tání: -90.6
Bod vzplanutí: -4
MDL Number: MFCD00009544
CAS číslo: 142-82-5
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR