Tisk…
Supelco® Ascentis® Express PFAS HPLC Columns
Dodavatel: Merck
53557-U 53564-U 53559-U 53560-U 53562-U 53563-U 53565-U 53570-U 53572-U 53573-U
53557-UEA 18080 CZK
53557-U 53564-U 53559-U 53560-U 53562-U 53563-U 53565-U 53570-U 53572-U 53573-U
Supelco® Ascentis® Express PFAS HPLC Columns
Kolony chromatografické
Ascentis® Express PFAS HPLC columns, with Fused-Core® technology delivers necessary retention to perform in EPA methods 537.1, 533 and 8327. Fast and high resolution LC-MS separation of PFAS.
  • 2,7 µm Fused-Core® particle for reliable and high efficiency separations and lower column back pressure compared to sub 2 particles
  • Excellent suitability for MS detection
  • Application-related lot analysis and single column performance testing
The U.S. Food and Drug Administration (FDA) and U.S. Environmental Protection Agency (EPA) have initiated actions againstMore Product Information