Tisk…
Soxhlet extractors
Dodavatel: Lenz Laborglas GmbH & CO.KG
05360025 05360031 05360037 05360043 05360046 05360049 05360058 05360070 05365025 05365031 05365037 05365043 05365049 05395125 05395131 05395137 05395143 05395149 05395158 05395170 05360052 05395146 05395152 05365046
533-2131EA 6990 CZK
533-2131 533-2132 533-2133 533-2134 533-2135 533-2136 533-2138 533-2139 533-0039 533-0040 533-0041 533-0042 533-0043 533-2141 533-2142 533-2143 533-2144 533-2146 533-2148 537-1000 533-2137 533-2145 533-2147 537-0030
Soxhlet extractors
Extraktory Extraktory Soxhlet
DURAN®, borosilicate glass. For extraction from solids with solvents.
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Informace o dodávce: The complete apparatus consists of a round bottom flask with NS 29/32 socket, an extractor with NS 29/32 cone and a condenser with GL 14 tubing connectors. All parts are available separately.