Tisk…
Conical separating funnels
Dodavatel: Lenz Laborglas GmbH & CO.KG
04000028 04000037 04000049 04000058 04000070 04000073 04002073 04004170 04002028 04002037 04002049 04002058 04002070 04004128 04004137 04004149 04004158 04004173 04006128 04006137 04006149 04006158 04006170 04006173
527-1131EA 1480 CZK
527-1131 527-1132 527-1133 527-1224 527-1134 527-1135 527-1226 527-1140 527-1221 527-1222 527-1223 527-1214 527-1225 527-1136 527-1137 527-1138 527-1139 527-1141 527-1201 527-1202 527-1203 527-1204 527-1205 527-1206
Conical separating funnels
Nálevky Nálevky dělící
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.