Tisk…
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Dodavatel: Corning
SCT-200-SS-O SCT-200-SS-G SC-LP-BR SCT-050-A-S SCT-050-C-S SCT-050-SS-A-S SCT-050-SS-G SCT-050-SS-R SCT-150-A-S SCT-150-C-S SCT-150-SS-C-S SCT-200-A-S SCT-200-C-S SCT-200-SS-C-S ST-150-C-S ST-200-C-S SCT-050-SS-C SCT-150-C SCT-150-SS-C SCT-200-C SCT-200-SS-C SCT-200-SS-A-S SCT-050-SS-B-S SCT-050-SS-C-S SCT-050-SS-G-S SCT-050-SS-R-S SCT-050-SS-W-S SCT-050-SS-Y-S SCT-150-SS-A-S SCT-150-LP-C SCT-45-C-S ST-050-SS-S ST-200-SS-S
212-0065EA 4700 CZK
212-0065 525-0899 525-0805 525-0326 525-0325 525-0340 525-0858 525-0863 525-0328 525-0327 525-0338 525-0342 525-0343 525-0339 212-0348 212-0353 525-0315 525-0317 525-0319 525-0321 525-0323 525-0298 AXYGSCT-050-SS-B-S 525-0337 525-0861 525-0864 525-0868 525-0870 525-0487 525-0878 525-0909 525-0913 525-0914
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Zkumavky Mikrozkumavky centrifugační
PP, sterile or non sterile, conical base with or without skirt.
  • Easily perforated screw cap with extra long rim and O-ring to ensure effective, uniform closure for centrifugation
  • The rims of the tubes and caps are ribbed for easier opening and closing with one hand
  • Certified DNase-, RNase- and endotoxin-free
Designed for many laboratory applications such as the collection, treatment, centrifugation, long-term storage and transport of inhibitor enzymes or other reagents.
Volume: 0,5, 1,5 and 2,0 ml.
Max. RCF: 20000×g.
Informace o dodávce: NonMore Product Information