Tisk…
Floor Standing Centrifuges and Packages, Megafuge ST4F and ST4FR Plus
Dodavatel: Thermo Scientific
75009961 75009948 75009962 75017963 75017961 75017962 75016089
521-3425EA 349270 CZK
521-3425 521-3426 521-3427 521-3451 521-3453 521-3454 521-3456
Floor Standing Centrifuges and Packages, Megafuge ST4F and ST4FR Plus
Centrifugy Centrifugy na zemi stojící
The Thermo Scientific™ Megafuge ST4F/ST4FR Plus floor-standing centrifuges offer intuitive controls and versatility for power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors.
Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotorMore Product Information