Tisk…
Microcentrifuges, high speed, universal, 5430 / 5430 R (General Lab Product)
Dodavatel: EPPENDORF
5428000.610 5427000.015 5427000.210 5427000.415 5427000.610 5428000.010 5428000.205 5428000.410 5427000.178
521-2647EA 214870 CZK
521-2647 521-4287 521-2634 521-2635 521-4288 521-2636 521-2646 521-2637 521-2785
Microcentrifuges, high speed, universal, 5430 / 5430 R (General Lab Product)
Centrifugy Mikrocentrifugy
These unique cross-over centrifuges combine the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.
  • Can be fitted with 12 different rotors for many applications (microtubes, PCR strips, cryotubes, conical tubes, PCR plates, deep-well plates, slides)
  • Five program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Soft-touch one-finger lid closure for ergonomic operation
  • Eppendorf QuickLock® – System for quickly opening and closing the rotor lid
  • Easy-to-use operator
More Product Information