Tisk…
Centrifuges, bench top, refrigerated, 5920 R (IVD marked)
Dodavatel: EPPENDORF
5948000.417 5948000.018 5948000.212 5948000.310 5948000.514 5948000.611 5948000.816 5948000.914
521-2561EA 461700 CZK
521-2561 521-2555 521-2557 521-2559 521-2563 521-2565 521-2569 521-2775
Centrifuges, bench top, refrigerated, 5920 R (IVD marked)
Centrifugy Centrifugy stolní
This centrifuge features a superior capacity of up to 4×1000 ml which makes it the ideal instrument for high-throughput and large volume applications. It has a powerful refrigeration system with advanced temperature management to keep samples safe.
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • FastTemp function for quick pre-cooling; FastTemp pro® function allows for automatic precooling based on pre-programmable time and date
  • Continuous cooling maintains a constant temperature after the run has been completed – samples
More Product Information