Tisk…
Rotor for Cryofuge™ 6000i/8500i series
Dodavatel: Thermo Scientific
75006606
521-1384EA 438120 CZK
521-1384
Rotor for Cryofuge™ 6000i/8500i series
Centrifugy Rotory
Windshielded rotor for the large capacity centrifuges Cryofuge 6000i and 8500i.
  • Swinging bucket
  • Rotor body made of stainless steel; bucket made of aluminium
  • Comes with a lid but without buckets