Tisk…
Centrifuges, benchtop, ventilated/refrigerated, ROTINA 420 / 420 R
Dodavatel: Hettich
4701 4706
521-1155EA 167810 CZK
521-1155 521-1156
Centrifuges, benchtop, ventilated/refrigerated, ROTINA 420 / 420 R
Centrifugy Centrifugy stolní
High performance, compact centrifuges developed for high sample throughput as well as for high sample volumes.
  • User friendly centrifuge with automatic lid locking, ergonomic and advanced control panel, 98 program storage spaces, easily exchangeable rotors
  • Metal housing and lid, stainless steel centrifuging chamber, viewing port in the lid
  • Safety features: Lid locking and holding during rotor run, emergency lid lock release, lid dropping protection, motor overheat protection, chamber overheat protection (ROTINA 420 R), imbalance switch off,
More Product Information