Tisk…
Filtry ventilační, PolyVENT™, Whatman™
Dodavatel: Whatman products (Cytiva)
6713-0425 6713-1075 6713-1650 6713-5036 6713-1651
516-4455EA 7050 CZK
516-4455 516-4456 516-4457 516-4459 WHAT6713-1651
Filtry ventilační, PolyVENT™, Whatman™
Filtry Filtry odvzdušňovací
Whatman PolyVENT disc filters are integral venting filters that work bidirectionally to prevent contamination from entering containers like fermentation vessels during incubation.
  • 0,2 µm hydrophobic PTFE air filters are ideal industrial air filter media
  • Validated for 50 steam autoclave cycles; compatible with ethylene oxide (EtO)
  • Testable by water break through (WBT) test or bubble point testing
  • Manufactured in clean room facilities
  • Filtration area 500, 1000, and 2000 cm² to support a range of venting applications
Draining or filling of incubators,More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Ø 25 mm 75 mm 50 mm 36 mm
Maximální tlak 2 bar 2 bar 2 bar 2 bar