Tisk…
Membrane filter funnels, PVDF, Microsart® @filter 100
Dodavatel: Sartorius
16D04-10-06-BL 16D04-10-06-TG 16D04-10-06ACG
516-1157EA 12460 CZK
516-1157 516-1158 516-1159
Membrane filter funnels, PVDF, Microsart® @filter 100
Nálevky Nálevky filtrační membránové
Ready to use, sterile Microsart® @filter units combine a funnel, a filter base and a membrane filter in one unit. They have been specifically developed for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics and other liquids.
  • Fully single-use design and sterile packaging help prevent secondary contamination
  • Click-fit closure for effortless funnel removal
  • Reliable results as no liquid remains after filtration
  • Transparent funnel for easy monitoring of the filtration progress
  • Filtration units with widest selection of integrated
More Product Information